Peristaltic pumps, DosiPump DP 500 and DosiPump DP 1000
Leverancier: BioTool
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
740000-UK 500501-UK 500501-CH 740000-CH 730000-UK 730000-EU 730000-CH 740000-EU
710-1562EA 4250 EUR
710-1562 710-1554 710-1552 710-1560 710-1559 710-1558 710-1557 710-1561
Peristaltic pumps, DosiPump DP 500 and DosiPump DP 1000
Pompen Peristaltische pompen
The DosiPump is an intelligent peristaltic pump which allows precise and reproducible dosing. Capable of single volume dosing and multiple dosing in both directions. The easy to use DosiPump provides versatility to undertake any application requiring fast, reproducible and precise dispensing of liquids. The DosiPump comes with a pump head which eliminates pulsation effects that are prevalent with many conventional peristaltic pumps. The result is a precise and repeatable liquid dose for volumes up to 10 L with a ±0,1 ml tolerance.
Available in the basic 500 model, or the 1000 model, withMore Product Information