Petri dish filling system, PetriSwiss PS20 mini
Leverancier: BioTool
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
800000.ECO-CH 810000-UK 810000-EU 810000-CH 810000.CO-UK 810000.CO-EU 810000.CO-CH 800000-UK 800000-EU 800000-CH 800000.ECO-UK 800000.ECO-EU 820000-CH
710-1671EA 11460 EUR
710-1671 BTOL810000-UK 710-1678 710-1677 BTOL810000.CO-UK 710-1676 710-1675 710-1707 710-1674 710-1673 BTOL800000.ECO-UK 710-1672 710-1679
Petri dish filling system, PetriSwiss PS20 mini
Petrischaalvullers
PetriSwiss PS 20 fills 20 dishes directly in the PetriRack. There is no need for manual stacking or any other manual operations. It is ideal for production of small, special-purpose agar batches. The dishes are safely stacked in the PetriRack and can even be transported, chilled or incubated in the rack. The rack can also be fully autoclaved. The dishes can be taken continuously from the rack. All program data, parameters and operating states are displayed clearly and understandably on a graphic display. The unit is very operator-friendly and is operated using a state-of-the-art touch screen.More Product Information