Sodium metatungstate monohydrate ≥85% WO₃ basis, purum p.a., Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate

71913-100G 71913-25G
71913-100GEA 280 EUR
71913-100G 71913-25G
Sodium metatungstate monohydrate ≥85% WO₃ basis, purum p.a., Sigma-Aldrich®
Sodium metatungstate monohydrate

Reagent for the preparation of heavy liquid, for sink-float analysis.


A new medium for density-gradient centrifugation, the density can be adjusted to up to 3.1 g/ml

Formule: Na₆O₃₉W₁₂·H₂O
MW: 2986.13 g/mol (2968.12 anhy)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061466
CAS nummer: 12141-67-2
EINECS: 412-770-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eb95f60df8a259a449623cad10a759e96ad60b6&vid=f6c50d2504b787beb1b5804aea58661c1a5498a9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR