Centrifugeerbuisjes

Leverancier: Simport Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T405-1 T405-33 T405-1A T406-1 T406-33 T406-1A T405-2 T405-2A T405-3 T405-6 T405-6A T406-2 T406-2A T406-3 T406-6 T406-6A
720-0556EA 285 EUR
720-0556 720-0561 720-0557 720-0564 720-0569 720-0565 720-0558 720-0559 720-0560 720-0562 720-0563 720-0566 720-0567 720-0568 720-0570 720-0571
Centrifugeerbuisjes
Buisjes Cultuurbuisjes
PP or PS, graduated, disposable, sterile.

  • White marking field
  • Gradation distinguishing 0,1 (4 ml tube) and 0,25 ml (12 ml tube)
  • Biological inert
  • Durable, reduces the risk in handling potentially hazardous material

Can be used for most routine laboratory procedures.

Resist temperatures from −190 to +120 °C (PP) and up to 70 °C (PS).

Max. RCF: 3000×g (PP); 1400×g (PS).

Leveringsinformatie: With or without cap available.
Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR