Tissue cassettes in QuickLoad™ taped stacks, Histosette® II, M492T
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M492-10T M492-11T M492-12T M492-2T M492-3T M492-4T M492-5T M492-6T M492-7T M492-8T M492-9T
720-1367EA 567 EUR
720-1367 720-1368 720-1369 720-1370 720-1371 720-1372 720-1373 720-1374 720-1375 720-1376 720-1377
Tissue cassettes in QuickLoad™ taped stacks, Histosette® II, M492T
Histologie-cassettes Cassettes voor weefselbehandeling
Disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage.
  • The anterior writing area is slanted at a 45° angle to make the cassette more suitable for use with certain types of cassette-labelling instruments
  • Available in non cytotoxic, non metallic colours
Moulded from a special high density acetal polymer, these patented cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximise fluid exchangeMore Product Information