Inbedcassetten, Macrosette®, M512
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M512
720-1627EA 489 EUR
720-1627
Inbedcassetten, Macrosette®, M512
Histologie-cassettes Cassettes voor weefselbehandeling
Acetylpolymeer.
  • Disposable plastic cassettes designed to hold larger tissue specimens - same size as Histosette® cassette but twice as deep (12 mm)
  • Highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Front face inclined 45° to be usable with leading automatic labelling systems
b×d×h: 41,1×28,5×13 mm
Verpakking: Elke verpakking bevat 3 dozen met 250 cassettes.
Order Now
Specificaties
Environmentally Preferable