Blood collection tubes with clot activator, vacuum, Vacutest®
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11020 11030 11002 11010 11005
720-2734EA 32.6 EUR
720-2734 720-2735 720-2730 720-2731 720-2763
Blood collection tubes with clot activator, vacuum, Vacutest®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in three sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
  • 18
More Product Information