N,N-Dimethylformamide, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

MQ100

72438-5ML-F
72438-5ML-FEA 177 EUR
72438-5ML-F
N,N-Dimethylformamide, Supelco®
N,N-Dimethylformamide
Formule: HCON(CH₃)₂
MW: 73.09 g/mol
Kookpunt: 153
Smeltpunt: –61 °C
Dichtheid: 0.949 g/cm³ (20 °C)
Vlampunt: 57.5 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003284
CAS nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
Bereik: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=816d104c2b515bc860ffb4b0138c3572ac2b0c2d&vid=1225866b521943c023c0b9bbeccc20c298e08f31&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR