SYPRO® Ruby Protein Gel Stain
Leverancier: LONZA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
50562 50564
730-1305EA 593 EUR
730-1305 730-1307
SYPRO® Ruby Protein Gel Stain
Kleurstoffen Kleuringen voor elektroforese
SYPRO™ Ruby Protein Gel Stain is highly sensitive, simple to use fluorescent stain that can accurately quantitate protein expression levels, and is compatible with standard fluorescent visualization systems and downstream identification techniques such as mass spectrometry.
  • Visualize proteins prior to transfer - Does not interfere with protein activity
  • Versatile - Excellent for staining native proteins
  • Safe - No acids or organic solvents necessary
  • Sensitive - Detects as little as 4-8 ng protein per band