DNA specimen collection cards, Ahlstrom-Munksjö Gensaver™ and GenSaver™ 2.0
Leverancier: Ahlstrom-Munksjö
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8.566.0000.B-N 8.566.0001.B-N 8.567.0000.B-N 8.567.0001.B-N 8.567.0002.B-N 8.566.0003.B-G 8.566.0003.B-K 8.566.0003.B-N 8.567.0003.B-G 8.567.0003.B-K 8.567.0003.B-N
730-0662EA 420 EUR
730-0662 730-0684 730-1349 730-1486 730-1487 BINZ8.566.0003.B-G BINZ8.566.0003.B-K BINZ8.566.0003.B-N BINZ8.567.0003.B-G BINZ8.567.0003.B-K BINZ8.567.0003.B-N
DNA specimen collection cards, Ahlstrom-Munksjö Gensaver™ and GenSaver™ 2.0
Opslagsystemen voor nucleïnezuren
GenSaver™ and GenSaver™ 2.0 collection cards are treated with a proprietary chemistry intended to prevent environmentally induced degradation. These cards allow long term preservation of DNA from biological fluids stored at ambient temperature.
  • Chemical treatment of these cards allows long-term ambient protection of DNA from dried matrix spots and improved qualitative and quantitative DNA recovery
  • Cards are manufactured in accordance to the world’s first international standard for forensic consumables, minimising the risk of human DNA contamination
  • DNA
More Product Information