Cap strips for Piko® UTW plates
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TCS-1080
731-0392EA 76 EUR
731-0392
Cap strips for Piko® UTW plates
Afdichtingen Afdichtingen van microtiterplaten
These flat cap strips provide an extremely tight seal in UTW (ultra-thin walled) PCR plates. They are an ideal choice for low volume reactions where it is important to minimize vapor loss.
  • 8-cap strip
  • Optically clear for fluorescence detection
  • Tight sealing for low-volume samples and consistent quantification results
  • Ideal for sealing 24-well Piko UTW PCR plates
The optically-clear flat caps provide great transmission for all fluorescence detection applications, such as qPCR.