Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030128.508 0030128.516 0030128.524 0030128.532 0030128.540 0030128.940 0030128.974 0030128.958 0030128.931 0030128.966 0030129.342 0030129.350 0030129.628 0030113.578
732-0101EA 227 EUR
732-0101 732-0102 732-0103 732-0104 732-0105 EPPE0030128.940 EPPE0030128.974 EPPE0030128.958 EPPE0030128.931 EPPE0030128.966 737-0183 737-0184 EPPE0030129.628 737-0257
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Microplaten PCR-platen
Eppendorf's twin.tec® 384-well, 45 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimising the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid and torque-resistant polycarbonate