RNA ScreenTape Analysis
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5067-5576 5067-5577 5067-5578
732-3997EA 208 EUR
732-3997 732-3998 732-3999
RNA ScreenTape Analysis
Elektroforesesystemen Systemen voor capillaire elektroforese
Reliable, efficient, and objective measurements of quality and quantity of RNA samples.
  • Utilises RINe - a user independent algorithm for accurate and objective assessment of eukaryotic or prokaryotic total RNA integrity
  • Provides fast results with data delivered in approximately 1 minute per sample
  • Minimal sample consumption is required as only 1 µl of sample is needed for accurate data
  • RNA sample detection down to 5 ng/µl is possible
  • Objective quality control of precious RNA samples is enabled within RNAseq, microarray analysis, and quantitative RT-PCR
More Product Information
Order Now
Specificaties
Environmentally Preferable