Protease inhibitor cocktail, Plant ProteaseArrest™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-332
733-1948EA 196 EUR
733-1948
Protease inhibitor cocktail, Plant ProteaseArrest™
Celcultuursupplementen en -additieven
A broad range, 100X concentrated, ready-to-use protease inhibitor cocktail, Plant ProteaseArrest™ inhibits plant serine, cysteine and other plant specific proteases including aminopeptidases, aspartic and metalloproteases.
  • Plant specific protease inhibitor cocktail
  • 100X concentrated, ready-to-use solution
Applications inhibition of protease activity in protein preparations of plant lysates and protection of proteins from proteolysis in such applications as electrophoresis, purification, storage and assays.