VWR®, DNA ladders

Leverancier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

733-2602EA 50.8 EUR
733-2602 733-2601 733-2603
VWR®, DNA ladders
Reagentia voor elektroforese Molecuulgewichtmerkers

VWR® DNA ladders are convenient dsDNA ladders supplied in a loading buffer, which are ready to use on agarose and SDS DNA gels.


  • Ready to use loading buffer
  • For direct loading and easy visualisation
  • Suitable with TBE and TAE electrophoresis systems
  • Clear, distinct bands
  • Blue dye front running at 100 to 300 bp at 0,5 to 1,5% agarose


Bestelinformatie: Supplied in 0,5 ml packs for 100 lanes.

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR