96-well microtestplaten, Falcon®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
353072 353075 353077 353227 353296 353910 353936 351172 351177 353916 353219 353376 353377
734-0023EA 192 EUR
734-0023 734-0025 734-0027 734-0057 734-0396 734-0080 736-2027 734-0954 734-0955 734-1376 734-2480 732-2194 732-2195
96-well microtestplaten, Falcon®
Microplaten Celcultuurplaten
Falcon® 96-well microplates are available in a variety of configurations and surface chemistries to meet a wide range of applications with fluorescence, luminescence, radiometric or colorimetric endpoints.
  • Flat bottom, with lid
  • Black wells with clear bottom
  • Working volume: 25 to 340 µl
  • Convenient, peel-open packaging
  • Available with untreated or tissue culture-treated surface chemistries
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
Corning® Primaria™ tissue culture treatment additionally incorporates nitrogen-containingMore Product Information
Order Now
Specificaties
Environmentally Preferable