Low volume microplates 96-well polystyrene, Corning® Stripwell™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2481 2483 2485
732-5577EA 853.61 EUR
732-5577 734-1202 734-1204
Low volume microplates 96-well polystyrene, Corning® Stripwell™
Microplaten Immunologie-microplaten en -strips
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an ‘egg-crate’ style strip holder that allows everyone well to be positioned back into the microplate once broken.
  • Stripwell microplates have 96-well flat bottom polystyrene format
  • Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
  • Strips 1×8 are designed to fit only one way into the strip holder, eliminating the chance of misorientation
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering RECs, ISO 14001 - Kennebunk