Microplates, 384-well, Whatman™ UNIFILTER®
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7700-1101 7700-2110
512-1141EA 2010 EUR
512-1141 734-1235
Microplates, 384-well, Whatman™ UNIFILTER®
Microplaten Filtreerplaten
Cytiva's Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
  • Usable with gel filtration media, allowing high-throughput removal of dye blobs
  • Long drip director prevents well-to-well cross contamination
  • Rigid, clear polystyrene microplate can withstand centrifugation
  • Membrane media option of DNA binding polymer or glass microfiber
Whatman UNIFILTER microplate options include membrane types andMore Product Information
Order Now
Specificaties
Environmentally Preferable