Centrifugebuisjes, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
430290 430291 430304 430828 430829 4558 431472
734-1827EA 233.08 EUR
734-1827 734-1828 734-1829 734-1868 734-1869 525-3414 525-0479
Centrifugebuisjes, Corning®
Buisjes Centrifugebuisjes
Corning 50 ml centrifuge tubes made of PP or PET.
  • Conical, with HDPE CentriStar™ or plug seal caps or without cap, sterile
  • Black printed graduations and a large white marking spot
  • Certified non pyrogenic and DNase/RNase-free
The CentriStar™ cap with leakproof design has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
Bestelinformatie: Schuim rekken en CentriStar™ doppen zijn afzonderlijk beschikbaar.
Verpakking: In bulk of in rekken.