ECM Mimetic celcultuurplaten, Corning® PureCoat™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
356270 356271
734-2494EA 339.34 EUR
734-2494 734-2495
ECM Mimetic celcultuurplaten, Corning® PureCoat™
Microplaten Celcultuurplaten
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor and primary cell types.
  • Available peptides: Human collagen I and human fibronectin
  • Delivers similar adhesion capabilities as the native protein,
More Product Information