Avantor® Small Scale Magnetic Mixing Systems
Leverancier: JM SEPARATIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
734-3666EA 37080 EUR
734-3666 734-3663 JMBVAFHM20MMS-0101 JMBVAFHM20MMS-0102
Avantor® Small Scale Magnetic Mixing Systems
Roerders Bovenroerders
The Avantor Small scale magnetic mixing system is a robust, flexible, high-quality, expert-designed solution for small volume single use mixing needs up to 20 L.
  • Intuitive software: Controlled by a user-friendly software program that runs on a 7" colour touchscreen display. This software allows users to save mixing profiles and customize their mixing settings
  • Integrated loadcell: The system comes with an integrated loadcell as standard that measures the weight of the vessel and contents, providing accurate feedback on the mixing process
  • Lexan support
More Product Information