Microplates, 96-well
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
655081_40 655001_40 655061_40 650061_40 651001_40 651061_40 655092_50 650001_40 675001_40 675061_40 655080_40 655001-000
391-3390EA 87 EUR
391-3390 736-0227 738-0015 738-0005 738-0008 738-0009 738-0167 738-0004 738-0032 738-0033 738-0017 GREI655001-000
Microplates, 96-well
Microplaten HTS-microplaten
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
  • Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
  • Free of DNase, RNase and human DNA, and are non-pyrogenic