Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030128.648 0030128.680 0030128.672 0030128.664 0030128.656 0030128.850 0030128.842 0030128.796 0030128.788 0030128.770 0030133.331 0030129.318 0030133.307 0030129.300 0030133.358 0030133.382 0030129.601 0030129.849 0030129.857 0030113.560 0030113.616 0030113.624 0030113.632
732-0107EA 153 EUR
732-0107 391-2054 732-0110 732-0109 732-0108 EPPE0030128.850 EPPE0030128.842 737-0164 EPPE0030128.788 EPPE0030128.770 731-0411 737-0180 731-0408 731-0414 731-0413 713-0021 EPPE0030129.601 EPPE0030129.849 EPPE0030129.857 737-0256 EPPE0030113.616 EPPE0030113.624 EPPE0030113.632
Eppendorf twin.tec® PCR Plates 96-well, 150 µl
Microplaten PCR-platen
Eppendorf's twin.tec® 96-well, 150 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. The SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid
More Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage Lage impact op de fabricage,Duurzame materialen
Milieu certificering Renewable Energy Certificate, ISO 14001 ISCC PLUS, Renewable Energy Certificate, ISO 14001