n-Pentaan 99,0 GC_ASSAY_METHOD, Puriss. p.a., Riedel-de Haën™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Pentane

76871-2.5L 76871-1L
76871-2.5LEA 320 EUR
76871-2.5L 76871-1L
n-Pentaan 99,0 GC_ASSAY_METHOD, Puriss. p.a., Riedel-de Haën™
n-Pentaan

Beilstein Number: 969132

Formule: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Kookpunt: 36…37 °C
Smeltpunt: –131 °C
Dichtheid: 0.631 g/cm³ (15 °C)
Vlampunt: <–35 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009498
CAS nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
Bereik: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.357 - 1.359
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.05 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Isopentane (GC) Max. 1.0 %
2-methylpentane (GC) Max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR