Titanium(II) oxide ≥99.5% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Titanium oxide, Titanium monoxide

77126.06 77126.14 77126.22 77126.36
77126.06EA 42.3 EUR
77126.06 77126.14 77126.22 77126.36
Titanium(II) oxide ≥99.5% (metals basis)
Titanium(II) oxide

Purity: 99.5% CAS No.: 12137-20-1 Molecular Formula: TiO Formula Weight: 63.90 Physical Form: -325 Mesh Powder Density: 4.95 Melting Point: 1700° Boiling Point: 3000° MDL No.: MFCD00036279

Formule: OTi
MW: 63.88 g/mol
Kookpunt: >3000 °C
Smeltpunt: 1700 °C
Dichtheid: 4.95
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036279
CAS nummer: 12137-20-1
EINECS: 235-236-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR