Titanium(IV) ethanolate (≥33% TiO₂)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Titanium (IV) ethoxide, Titanium tetraethanolate, Tetraethyl orthotitanate

77142.06 77142.18 77142.30
77142.06EA 35 EUR
77142.06 77142.18 77142.30
Titanium(IV) ethanolate (≥33% TiO₂)
Titanium(IV) ethanolate

CAS No.: 3087-36-3 Molecular Formula: C8H20O4Ti Formula Weight: 228.15 Flash Point: 28°(84°F) Physical Form: Liquid Density: 1.088 Refractive Index: 1.5043 Sensitivity: Moisture Sensitive Appearance: Pale yellow Odour: Alcohol-like Melting Point: -40° Boiling Point: 150-152°/10mm Solubility: Soluble in organic solvents. Decomposes in water UN No.: 2924 MDL No.: MFCD00009071

Formule: C₈H₂₀O₄Ti
MW: 228.12 g/mol
Kookpunt: 150…152 °C (10 mmHg)
Smeltpunt: –40 °C
Dichtheid: 1.088
Vlampunt: 28 °C (84 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009071
CAS nummer: 3087-36-3
EINECS: 221-410-8
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR