Supelco® PESTANAL® Organic Reference Standard, Mepiquat iodide-(methyl-d₃)

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

78278-10MG
78278-10MGEA 554 EUR
78278-10MG
Supelco® PESTANAL® Organic Reference Standard, Mepiquat iodide-(methyl-d₃)
Standaarden Organische standaarden

  • GHS signal: GHS07


Opgelet: P264 - P270 - P280 - P301 + P312 - P305 + P351 + P338 - P337 + P313

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67b304e7cce0a108eb58fd9734b018eae963dbbc&vid=67b304e7cce0a108eb58fd9734b018eae963dbbc&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable
Synonym(s) 1-Methyl-1-methyl-d₃-piperidinium iodide
M-Clarity quality level MQ100
CAS No. 32317-85-4
MDL No. MFCD00032083
Moleculair gewicht 244,13
Beschrijving van de gevarenklasse H302 - H319
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR