Water TOC, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Hydrogen oxide

78533-1L
78533-1LEA 46 EUR
78533-1L
Water TOC, Supelco®
Water
Formule: H₂O
MW: 18.02 g/mol
Kookpunt: 100
Smeltpunt: 0
Dichtheid: 1 g/cm³ (4 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011332
CAS nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76327ff668c173af36615fd453223d4004b161d2&vid=873b85b9ab3b3447a9945074449a37be1cbd90fb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR