Detergent removal system, DetergentOUT™ Tween®
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-214 786-215
786-214EA 478 EUR
786-214 786-215
Detergent removal system, DetergentOUT™ Tween®
Eiwitextractie Reagentia voor eiwitextractie
DetergentOUT™ Tween® is a spin column format detergent removal resin for polysorbate or Tween® detergents or surfactants.
DetergentOUT™ Tween® specifically removes polysorbate detergents without significant loss of proteins, dilution of the protein solution, or change to the buffer composition of the protein solution.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d52c1043214149edc1c697a3ffffe58f6c025f&vid=0633c24839d746937941c948790c95906898733e&prid=web&clid=SAPDAM