Nonidet® P 40 Substitute (NP-40)

Leverancier: G Bioscienes
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

786-511 786-512
786-511EA 489 EUR
786-511 786-512
Nonidet® P 40 Substitute (NP-40)
Nonidet® P 40 Substitute (NP-40)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00132851
CAS nummer: 9016-45-9
EINECS: 500-024-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR