Protein purification kits, HOOK™
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
786-628 786-630 786-640 786-641 786-629 786-631 786-634 786-632 786-635 786-642 786-643 786-633
786-628EA 919 EUR
786-628 786-630 786-640 786-641 786-629 786-631 786-634 786-632 786-635 786-642 786-643 786-633
Protein purification kits, HOOK™
Eiwitzuivering Protein Purification Kits
For the rapid purification of soluble, 6X His or GST tagged proteins.
  • For bacteria or yeast cells
  • Includes lysis buffer and lytic enzymes
  • Gravity or spin format
  • Nickel or cobalt chelating resin for 6X His
  • High binding glutathione resin for GST
HOOK™ protein purification kits allow for the purification of soluble protein from bacterial and yeast cultures. The kits are supplied with a specific PE LB™ lysis buffer and lytic enzyme (LongLife™ PE LB™ Lysozyme or LongLife™ Zymolyase®) for efficient cell lysis with maintenance of the tagged protein’s properties. The 6X His tagged proteins are then purified with high binding efficiency nickel chelating or cobalt chelating resin. Cobalt chelating resin has a lower binding affinity for 6X His tags, compared to nickel chelating resin, which results in less non specific binding and may result in slightly lower yields. The GST tagged proteins are purified with a high capacity glutathione resin.
HOOK™ protein purification kits are supplied in a gravity flow format for yields of 10 mg/250 ml of culture and spin kits for rapid elution of protein for ~1 mg/50 ml of culture.