Nitric acid/Rubidium concentrate 100 mM HNO₃ in H₂O, 1000 μg/L Rb in H₂O (from RbNO₃ salt)

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Nitric acid solution

78737-1L 78737-2.5L
78737-1LEA 99.3 EUR
78737-1L 78737-2.5L
Nitric acid/Rubidium concentrate 100 mM HNO₃ in H₂O, 1000 μg/L Rb in H₂O (from RbNO₃ salt)
Salpeterzuur
Formule: HNO₃
MW: 63.01 g/mol
Kookpunt: 87
Smeltpunt: -42
Dichtheid: 1.51
MDL Nummer: MFCD00011349
CAS nummer: 7697-37-2
Merck Index: 13,06610

Order Now

Specification Test Results

Solubility 20 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR