Natrium-diwaterstoffosfaat-monohydraat, granular, AR® ACS, Macron Fine Chemicals™

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

7892-04EA 0 EUR
7892-04 7892-06
Natrium-diwaterstoffosfaat-monohydraat, granular, AR® ACS, Macron Fine Chemicals™
Natrium-diwaterstoffosfaat-monohydraat
Formule: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Smeltpunt: 100
Dichtheid: 2.04 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149208
CAS nummer: 10049-21-5
EINECS: 231-449-2
Bereik: 01-2119489796-13
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

ACS - Appearance (small, white granules) Passes Test
ACS - Assay (NaH2PO4·H2O) 98.0 - 102.0 %
ACS - Calcium (Ca) ≤ 0.005 %
ACS - Chloride (Cl) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 0.001 %
ACS - Insoluble Matter ≤ 0.01 %
ACS - Iron (Fe) ≤ 0.001 %
ACS - pH of 5% Solution at 25°C 4.1 - 4.5
ACS - Potassium (K) ≤ 0.01 %
ACS - Solubility Passes Test
ACS - Sulfate (SO4) ≤ 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR