Natriumsulfaat, watervrij ≥99%, Reagent Grade, free-flowing, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

793531-12KG 793531-5KG
793531-12KGEA 291 EUR
793531-12KG 793531-5KG
Natriumsulfaat, watervrij ≥99%, Reagent Grade, free-flowing, Fluka™
Natriumsulfaat

  • ≥95% for general laboratory use.

Formule: Na₂SO₄
Smeltpunt: 884 °C
Dichtheid: 2,68 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003504
CAS nummer: 7757-82-6
EINECS: 231-820-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Solution 10 g to 100 mL (H₂O) complying
Loss on drying (130°C) Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR