3-Aminopropan-1-ol 99,0 GC_ASSAY_METHOD zS, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 3-Amino-1-propanol

MQ200

8.00423.0005 8.00423.0250 8.00423.9025
8.00423.0005EA 17.4 EUR
8.00423.0005 8.00423.0250 8.00423.9025
3-Aminopropan-1-ol 99,0 GC_ASSAY_METHOD zS, Sigma-Aldrich®
3-Aminopropan-1-ol

Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Formule: C₃H₉NO
MW: 75.11 g/mol
Kookpunt: 187
Smeltpunt: 11
Dichtheid: 0.982 g/cm³ (20 °C)
Vlampunt: 101
MDL Nummer: MFCD00008223
CAS nummer: 156-87-6
EINECS: 205-864-4
UN: 2735
ADR: 8 II

Order Now

Specification Test Results

Assay (GC, area%) ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C) 0.986 - 0.988
Water (K. F.) ≤ 0.30 %
Identity (IR) passes test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR