Benzenesulphonic acid for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.00468.0005 8.00468.0250 8.00468.1000 8.00468.9050 8.00468.9025 8.00468.1001
8.00468.0005EA 30.3 EUR
8.00468.0005 8.00468.0250 8.00468.1000 8.00468.9050 8.00468.9025 8.00468.1001
Benzenesulphonic acid for synthesis, Sigma-Aldrich®
Benzenesulphonic acid
Formule: C₆H₆O₃S
MW: 158.18 g/mol
Kookpunt: 319
Smeltpunt: 50
Dichtheid: 1.32 g/cm³ (47 °C)
Vlampunt: >100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011689
CAS nummer: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8 III
Merck Index: 13,01070

Order Now

Specification Test Results

Solubility 930 g/l (25 °C)
Melting Point 43 °C
Molar Mass 158.17 g/mol
Boiling Point 171 °C (0.13 hPa)
Vapor Pressure (20 °C)
Flash Point 100 °C
Density 1.32 g/cm3 (47 °C)
pH 2 (H2O, 20 °C) (saturated solution)
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR