Barnsteenzuuranhydride for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Dihydrofuran-2,5-dione

M-Clarity™ quality level = MQ200

8.00683.0100 8.00683.0500 8.00683.1000 8.00683.9050
8.00683.0100EA 38.2 EUR
8.00683.0100 8.00683.0500 8.00683.1000 8.00683.9050
Barnsteenzuuranhydride for synthesis, Sigma-Aldrich®
Barnsteenzuuranhydride
Formule: C₄H₄O₃
Kookpunt: 261 °C (1013 hPa)
Smeltpunt: 120 °C
Dichtheid: 1,503 g/cm³ (20 °C)
Vlampunt: 157 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005525
CAS nummer: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8,II
Merck Index: 12,09039

Order Now

Specification Test Results

Solubility 67 g/l (20 °C) Hydrolysis
Melting Point 116 - 120 °C
Molar Mass 100.07 g/mol
Bulk Density 550 - 600 kg/m3
Boiling Point 261 °C (1013 hPa)
Vapor Pressure 1.3 hPa (92 °C)
Flash Point 157 °C
Density 1.48 g/cm3 (20 °C)
pH 2.7 (20 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR