Ethyl vinyl ether stabilised, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Stabilisatie: Stabilised with potassium hydroxide
M-Clarity™ quality level = MQ200

8.01391.0100 8.01391.1000 8.01391.2500 8.01391.9040
8.01391.0100EA 35.8 EUR
8.01391.0100 8.01391.1000 8.01391.2500 8.01391.9040
Ethyl vinyl ether stabilised, Sigma-Aldrich®
Ethyl vinyl ether
Formule: C₄H₈O
MW: 72.11 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009248
CAS nummer: 109-92-2
EINECS: 203-718-4
UN: 1302
ADR: 3 I

Order Now

Specification Test Results

Solubility 8.3 g/l (15 °C)
Melting Point -115 °C
Molar Mass 72.11 g/mol
Boiling Point 36 °C (1013 hPa)
Vapor Pressure 560 hPa (20 °C)
Flash Point -45 °C
Refractive Index 1.3757 (20 °C, 589 nm)
Explosion Limit 1.3 - 12.0 %(V)
Density 0.75 g/cm3 (20 °C)
Ignition Point 178 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR