2-Amino-2-methyl-1,3-propanediol, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

8.01464.0050 8.01464.0250
8.01464.0050EA 55.3 EUR
8.01464.0050 8.01464.0250
2-Amino-2-methyl-1,3-propanediol, Sigma-Aldrich®
2-Amino-2-methyl-1,3-propanediol
Formule: C₄H₁₁NO₂
MW: 105.14 g/mol
Kookpunt: 272
Smeltpunt: 102…105 °C
Dichtheid: 1.211 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004678
CAS nummer: 115-69-5
EINECS: 204-100-7
UN: 3259
ADR: 8 III
Merck Index: 13,00447

Order Now

Specification Test Results

Solubility 2500 g/l (20 °C)
Melting Point 102 - 105 °C
Molar Mass 105.13 g/mol
Boiling Point 150 - 152 °C (13 hPa)
Vapor Pressure 13.3 hPa (152 °C)
Flash Point 100 °C
pH 10 - 11 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR