Butylamine for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 1-Butylamine, 1-Aminobutane, n-Butylamine

M-Clarity™ quality level = MQ200

8.01539.1000 8.01539.2500
8.01539.1000EA 50.3 EUR
8.01539.1000 8.01539.2500
Butylamine for synthesis, Sigma-Aldrich®
Butylamine
Formule: H₃C(CH₂)₂CH₂NH₂
MW: 73.14 g/mol
Kookpunt: 77
Smeltpunt: –50 °C
Dichtheid: 0.741 g/cm³ (20 °C)
Vlampunt: –8 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011690
CAS nummer: 109-73-9
EINECS: 203-699-2
UN: 1125
ADR: 3 (8) II
Merck Index: 12,01578

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -50 °C
Molar Mass 73.14 g/mol
Boiling Point 76 - 78 °C
Vapor Pressure 93 hPa (20 °C)
Flash Point -8.5 °C DIN 51755 Part 1
Refractive Index 1.4015 (20 °C, 589 nm)
Explosion Limit 1.5 - 9.8 %(V)
Density 0.74 g/cm3 (20 °C)
pH 12.6 (100 g/l, H2O, 20 °C)
Ignition Point 290 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR