2-Butanethiol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.01818.0005 8.01818.0250
8.01818.0005EA 27.8 EUR
8.01818.0005 8.01818.0250
2-Butanethiol for synthesis, Sigma-Aldrich®
2-Butanethiol
Formule: C₄H₁₀S
Kookpunt: 85 °C (1013 hPa)
Smeltpunt: –165 °C
Dichtheid: 0,829 g/cm³ (20 °C)
Vlampunt: –23 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004865
CAS nummer: 513-53-1
EINECS: 208-165-2
UN: 2347
ADR: 3,II
Merck Index: 12,01612

Order Now

Specification Test Results

Solubility 1.32 g/l (20 °C)
Melting Point -165 °C
Molar Mass 90.18 g/mol
Boiling Point 82 - 88 °C (1013 hPa)
Vapor Pressure 108 hPa (25 °C)
Flash Point -23 °C
Explosion Limit 1.7 - 9.6 %(V)
Density 0.83 g/cm3 (20 °C)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR