Broomazijnzuur ≥98.0% (by GC) for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.02260.0005 8.02260.0250 8.02260.1000
8.02260.0005EA 25 EUR
8.02260.0005 8.02260.0250 8.02260.1000
Broomazijnzuur ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Broomazijnzuur
Formule: BrCH₂COOH
Kookpunt: 206 °C (1013 hPa)
Smeltpunt: 46…49 °C
Dichtheid: 1,9335 g/cm³ (20 °C)
Vlampunt: >112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002678
CAS nummer: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 13,01384

Order Now

Specification Test Results

Solubility 93.8 g/l (25 °C)
Melting Point 46 - 49 °C
Molar Mass 138.94 g/mol
Boiling Point 208 °C (1013 hPa)
Vapor Pressure 0.158 hPa (25 °C)
Flash Point 112 °C
Density 1.93 g/cm3 (20 °C)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR