Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether

M-Clarity™ quality level = MQ200

8.02932.1000
8.02932.1000EA 132 EUR
8.02932.1000
Bis(2-ethoxyethyl)ether for synthesis, Sigma-Aldrich®
Bis(2-ethoxyethyl)ether
Formule: C₈H₁₈O₃
MW: 162.23 g/mol
Kookpunt: 189
Smeltpunt: –44 °C
Dichtheid: 0.91 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009254
CAS nummer: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -44 °C
Molar Mass 162.22 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 71 °C
Refractive Index 1.4115 (20 °C, 589 nm)
Density 0.91 g/cm3 (20 °C)
Ignition Point 174 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR