(±)-Epichloorhydrine for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.03296.0100 8.03296.0500 8.03296.1000 8.03296.9060 8.03296.9030 8.03296.8000
8.03296.0100EA 51.5 EUR
8.03296.0100 8.03296.0500 8.03296.1000 8.03296.9060 8.03296.9030 8.03296.8000
(±)-Epichloorhydrine for synthesis, Sigma-Aldrich®
(±)-Epichloorhydrine
Formule: C₃H₅ClO
Kookpunt: 117,9 °C (1013 hPa)
Dichtheid: 0,9985 g/cm³ (20 °C)
Vlampunt: 28 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005132
CAS nummer: 106-89-8
EINECS: 203-439-8
UN: 2023
ADR: 6.1 (3),II
Merck Index: 13,03642

Order Now

Specification Test Results

Solubility 60 g/l (20 °C)
Melting Point -57.2 °C
Molar Mass 92.52 g/mol
Boiling Point 116.5 °C (1013 hPa)
Vapor Pressure 16.5 hPa (20 °C)
Flash Point 28 °C
Refractive Index 1.4375 (20 °C, 589 nm)
Explosion Limit 2.3 - 34.4 %(V)
Density 1.18 g/cm3 (20 °C)
Ignition Point 416 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR