1,3-Diethyl-2-thiourea, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: N,N'-Diethylthiourea

M-Clarity™ quality level = MQ200

8.03520.0250 8.03520.5000 8.03520.0050
8.03520.0250EA 34.2 EUR
8.03520.0250 8.03520.5000 8.03520.0050
1,3-Diethyl-2-thiourea, Sigma-Aldrich®
1,3-Diethyl-2-thiourea
Formule: C₅H₁₂N₂S
MW: 132.23 g/mol
Kookpunt: 169
Smeltpunt: 75…78 °C
Dichtheid: 1.118 g/cm³ (20 °C)
Vlampunt: 186
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004925
CAS nummer: 105-55-5
EINECS: 203-308-5

Order Now

Specification Test Results

Solubility 38 g/l (20 °C)
Melting Point 75 - 78 °C
Molar Mass 132.22 g/mol
Bulk Density 735 kg/m3
Flash Point 186 °C
Density 1.12 g/cm3 (20 °C)
pH 7 (38 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR