1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: DBN, 2,3,4,6,7,8-Hexahydropyrrolo[1,2-a]pyrimidine, 1,5-Diazabicyclo[4.3.0]-5-nonene

M-Clarity™ quality level = MQ200

8.03603.0025 8.03603.0100
8.03603.0025EA 93.4 EUR
8.03603.0025 8.03603.0100
1,5-Diazobicyclo[4.3.0]-5-nonene for synthesis, Sigma-Aldrich®
1,5-Diazobicyclo[4.3.0]-5-nonene
Formule: C₇H₁₂N₂
MW: 124.19 g/mol
Kookpunt: 110
Dichtheid: 1 g/cm³ (20 °C)
Vlampunt: 94
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005554
CAS nummer: 3001-72-7
EINECS: 221-087-3
UN: 3267
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 124.18 g/mol
Boiling Point 110 °C (20 hPa)
Flash Point 94 °C
Refractive Index 1.519 (20 °C, 589 nm)
Density 1.04 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR