(±)-4-Methyl-2-pentanol for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: (±)-Methyl isobutyl carbinol

M-Clarity™ quality level = MQ200

8.05804.0100 8.05804.1000
8.05804.0100EA 38.2 EUR
8.05804.0100 8.05804.1000
(±)-4-Methyl-2-pentanol for synthesis, Sigma-Aldrich®
(±)-4-Methyl-2-pentanol
Formule: C₆H₁₄O
MW: 102.18 g/mol
Kookpunt: 132
Smeltpunt: –90 °C
Dichtheid: 0.8093 g/cm³ (20 °C)
Vlampunt: 54
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004550
CAS nummer: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3 III

Order Now

Specification Test Results

Solubility 16.4 g/l (20 °C)
Melting Point -60 °C
Molar Mass 102.18 g/mol
Boiling Point 130 - 133 °C (1013 hPa)
Vapor Pressure 7.06 hPa (25 °C)
Flash Point 41 °C
Explosion Limit 1 - 5.5 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 305 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR