3-Buten-2-one stabilised for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Stabilisatie: Stabilised with hydroquinone (HQ)
M-Clarity™ quality level = MQ200

8.05901.0100 8.05901.0250 8.05901.9040 8.05901.9030
8.05901.0100EA 58.03 EUR
8.05901.0100 8.05901.0250 8.05901.9040 8.05901.9030
3-Buten-2-one stabilised for synthesis, Sigma-Aldrich®
3-Buten-2-one
Formule: C₄H₆O
Kookpunt: 81,4 °C (1013 hPa)
Smeltpunt: –7 °C
Dichtheid: 0,8636 g/cm³ (20 °C)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00008777
CAS nummer: 78-94-4
EINECS: 201-160-6
UN: 1251
ADR: 6.1 (3, 8),I
Merck Index: 12,06214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c0b910689fbbbc834d57744b46744430d66145e&vid=accba25777bad1758e027949bef9960ba106c529&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR