3-Chloor-2-methylpropeen for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


M-Clarity™ quality level = MQ200

8.05905.0100 8.05905.0500
8.05905.0100EA 32.8 EUR
8.05905.0100 8.05905.0500
3-Chloor-2-methylpropeen for synthesis, Sigma-Aldrich®
3-Chloor-2-methylpropeen
Formule: C₄H₇Cl
MW: 90.55 g/mol
Kookpunt: 72
Smeltpunt: –80 °C
Dichtheid: 0.93 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000953
CAS nummer: 563-47-3
EINECS: 209-251-2
UN: 2554
ADR: 3 II
Merck Index: 13,02167

Order Now

Specification Test Results

Solubility 0.5 g/l (20 °C)
Melting Point -80 °C
Molar Mass 90.55 g/mol
Boiling Point 72 - 74 °C
Vapor Pressure 140 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4272 (20 °C, 589 nm)
Explosion Limit 2.2 - 10.4 %(V)
Density 0.93 g/cm3 (20 °C)
Ignition Point 540 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR