Sodium hydroxymethanesulphinate dihydrate ≥97.0% stabilised for synthesis, Sigma-Aldrich®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Sodium hydroxymethanesulphinate dihydrate(Chunks or pellets), Hydroxymethanesulphinic acid sodium salt dihydrate, Formaldehyde sodium sulphoxylate dihydrate, Sodium formaldehydesulphoxylate hydrate, Rongalite dihydrate, Sodium formaldehydesulphoxylate dihydrate

Stabilisatie: Stabilised with sodium carbonate (Na₂CO₃)
M-Clarity™ quality level = MQ200

8.06455.0100 8.06455.1000 8.06455.5000 8.06455.9050 8.06455.9025
8.06455.0100EA 30.5 EUR
8.06455.0100 8.06455.1000 8.06455.5000 8.06455.9050 8.06455.9025
Sodium hydroxymethanesulphinate dihydrate ≥97.0% stabilised for synthesis, Sigma-Aldrich®
Sodium hydroxymethanesulphinate dihydrate

This material contains approx. 5% sulfit resulting from the production process

Formule: CH₃NaO₃S·2H₂O
Smeltpunt: 63 °C
Dichtheid: 1,8 g/cm3 (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150598
CAS nummer: 6035-47-8
EINECS: 205-739-4

Order Now

Specification Test Results

Assay (iodometric, calc. as dihydrate) ≥ 97.0 %
Iron (Fe) ≤ 0.003 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR